siRNA Details
| Gene Symbol: LOC690326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109578.1 | NP_001103048.1 | 5 | 299 | ||
| XM_006237459.1 | XP_006237521.1 | 6 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690326| Gene Symbol: LOC690326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109578.1 | NP_001103048.1 | 5 | 299 | ||
| XM_006237459.1 | XP_006237521.1 | 6 | 473 | |||