siRNA Details
| Gene Symbol: Twf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135238.1 | NP_001128710.1 | 6 | 742 | ||
| GenBank mRNA | BC158614.1 | - | 6 | 690 | 7386008 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Twf2| Gene Symbol: Twf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135238.1 | NP_001128710.1 | 6 | 742 | ||
| GenBank mRNA | BC158614.1 | - | 6 | 690 | 7386008 | |