siRNA Details
| Gene Symbol: Ddx60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771220.1 | XP_008769442.1 | 14 | 2263 | ||
| Gene Symbol: LOC100911190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771241.1 | XP_008769463.1 | 18 | 3703 | ||

Ddx60
Genomic Map