siRNA Details
| Gene Symbol: Vom2r29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099496.1 | NP_001092966.1 | 3 | 734 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r29| Gene Symbol: Vom2r29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099496.1 | NP_001092966.1 | 3 | 734 | ||
Set Membership: |
|