siRNA Details
| Gene Symbol: LOC684403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770643.1 | XP_008768865.1 | 1 | 107 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC684403| Gene Symbol: LOC684403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770643.1 | XP_008768865.1 | 1 | 107 | ||