siRNA Details
| Gene Symbol: LOC686042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250422.2 | XP_006250484.1 | 1 | 204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC686042| Gene Symbol: LOC686042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250422.2 | XP_006250484.1 | 1 | 204 | ||