siRNA Details
| Gene Symbol: LOC686031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109490.1 | NP_001102960.1 | 1 | 365 | ||
| Gene Symbol: LOC686042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250422.2 | XP_006250484.1 | 1 | 400 | ||

LOC686031
Genomic Map