siRNA Details
| Gene Symbol: Rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127597.2 | NP_001121069.2 | 6 | 1323 | ||
| GenBank mRNA | BC166869.1 | - | 6 | 1323 | 9030307 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf168| Gene Symbol: Rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127597.2 | NP_001121069.2 | 6 | 1323 | ||
| GenBank mRNA | BC166869.1 | - | 6 | 1323 | 9030307 | |