siRNA Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750123.2 | XP_003750171.1 | 6 | 856 | ||
| XM_008764545.1 | XP_008762767.1 | 6 | 946 | |||
| XM_008764546.1 | XP_008762768.1 | 6 | 922 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpo| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750123.2 | XP_003750171.1 | 6 | 856 | ||
| XM_008764545.1 | XP_008762767.1 | 6 | 946 | |||
| XM_008764546.1 | XP_008762768.1 | 6 | 922 | |||