siRNA Details
| Gene Symbol: Tlr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101009.1 | NP_001094479.1 | 2 | 395 | ||
| GenBank mRNA | EF032638.1 | - | 2 | 395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlr8| Gene Symbol: Tlr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101009.1 | NP_001094479.1 | 2 | 395 | ||
| GenBank mRNA | EF032638.1 | - | 2 | 395 | ||