siRNA Details
| Gene Symbol: Tm6sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127654.1 | NP_001121126.1 | 7 | 674 | ||
| GenBank mRNA | BC158807.1 | - | 7 | 674 | 7324580 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm6sf2| Gene Symbol: Tm6sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127654.1 | NP_001121126.1 | 7 | 674 | ||
| GenBank mRNA | BC158807.1 | - | 7 | 674 | 7324580 | |