siRNA Details
| Gene Symbol: Mthfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109398.1 | NP_001102868.1 | 8 | 1048 | ||
| GenBank mRNA | FQ231869.1 | - | 4 | 359 | ||
| FQ234580.1 | - | 9 | 1100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mthfd2| Gene Symbol: Mthfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109398.1 | NP_001102868.1 | 8 | 1048 | ||
| GenBank mRNA | FQ231869.1 | - | 4 | 359 | ||
| FQ234580.1 | - | 9 | 1100 | |||