siRNA Details
Gene Symbol: Mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109398.1 | NP_001102868.1 | 7 | 937 | ||
GenBank mRNA | FQ231869.1 | - | 3 | 248 | ||
FQ234580.1 | - | 8 | 989 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109398.1 | NP_001102868.1 | 7 | 937 | ||
GenBank mRNA | FQ231869.1 | - | 3 | 248 | ||
FQ234580.1 | - | 8 | 989 |