siRNA Details
| Gene Symbol: TMEM63A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014698.2 | NP_055513.2 | 8, 9 | 1191 | ||
| XM_011544328.2 | XP_011542630.1 | 8, 9 | 1043 | |||
| XM_011544329.2 | XP_011542631.1 | 8, 9 | 2310 | |||
| XM_011544330.2 | XP_011542632.1 | 7, 8 | 1000 | |||
| XM_011544331.2 | XP_011542633.1 | 8, 9 | 874 | |||
| GenBank mRNA | AB018335.1 | - | 8, 9 | 819 | ||
| AK022502.1 | - | 8, 9 | 840 | |||
| AK222934.1 | - | 8, 9 | 821 | |||
| AK222944.1 | - | 8, 9 | 821 | |||
| BC030245.1 | - | 8, 9 | 824 | 5216414 | ||

TMEM63A
Genomic Map