siRNA Details
| Gene Symbol: Echdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101010.1 | NP_001094480.1 | 4 | 803 | ||
| GenBank mRNA | BC101897.1 | - | 4 | 803 | 7932594 | |
| FQ219501.1 | - | 4 | 842 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Echdc3| Gene Symbol: Echdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101010.1 | NP_001094480.1 | 4 | 803 | ||
| GenBank mRNA | BC101897.1 | - | 4 | 803 | 7932594 | |
| FQ219501.1 | - | 4 | 842 | |||