siRNA Details
| Gene Symbol: Pla2g2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109587.1 | NP_001103057.1 | 3 | 316 | ||
| XM_008764289.1 | XP_008762511.1 | 4 | 741 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g2f| Gene Symbol: Pla2g2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109587.1 | NP_001103057.1 | 3 | 316 | ||
| XM_008764289.1 | XP_008762511.1 | 4 | 741 | |||