siRNA Details
Gene Symbol: Pla2g2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109587.1 | NP_001103057.1 | 1 | 169 | ||
XM_008764289.1 | XP_008762511.1 | 2 | 594 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pla2g2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109587.1 | NP_001103057.1 | 1 | 169 | ||
XM_008764289.1 | XP_008762511.1 | 2 | 594 |