siRNA Details
| Gene Symbol: Zc3h10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191090.1 | NP_001178019.1 | 3 | 1412 | ||
| XM_008765041.1 | XP_008763263.1 | 3 | 1615 | |||
| XM_008765042.1 | XP_008763264.1 | 4 | 1876 | |||
| XM_008765043.1 | XP_008763265.1 | 4 | 1779 | |||

Zc3h10
Genomic Map