siRNA Details
| Gene Symbol: ZNF646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014699.3 | NP_055514.3 | 2 | 2544 | ||
| XM_005255710.3 | XP_005255767.1 | 2 | 3323 | |||
| XM_005255711.3 | XP_005255768.1 | 2 | 3282 | |||
| XM_005255712.3 | XP_005255769.1 | 2 | 3323 | |||
| XM_011545990.2 | XP_011544292.1 | 2 | 2549 | |||
| GenBank mRNA | AB002294.1 | - | 1 | 2678 | ||
| AK304808.1 | - | 2 | 2541 | |||
| BC035589.1 | - | 2 | 2530 | 5532753 | ||

ZNF646
Genomic Map