siRNA Details
| Gene Symbol: Fam154a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238373.2 | XP_006238435.1 | 2 | 414 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam154a| Gene Symbol: Fam154a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238373.2 | XP_006238435.1 | 2 | 414 | ||