siRNA Details
| Gene Symbol: Slc25a42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127590.1 | NP_001121062.1 | 5 | 352 | ||
| XM_006252950.2 | XP_006253012.1 | 5 | 433 | |||
| XM_006252951.2 | XP_006253013.1 | 5 | 433 | |||
| XM_008771155.1 | XP_008769377.1 | 5 | 950 | |||
| GenBank mRNA | BC160887.1 | - | 5 | 352 | 7387602 | |

Slc25a42
Genomic Map