siRNA Details
Gene Symbol: Hist1h4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771676.1 | XP_008769898.1 | 1 | 453 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hist1h4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771676.1 | XP_008769898.1 | 1 | 453 |