siRNA Details
| Gene Symbol: KIAA0408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014702.4 | NP_055517.3 | 5 | 1795 | ||
| GenBank mRNA | AB007868.1 | - | 3 | 2188 | ||
| AK091424.1 | - | 6 | 2151 | |||
| AK125983.1 | - | 6 | 2304 | |||
| BC018396.1 | - | 5 | 1633 | 4184033 | ||
| Gene Symbol: SOGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012279.2 | NP_001012279.1 | 11 | 5489 | ||

KIAA0408
Genomic Map