siRNA Details
Gene Symbol: Gje1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197092.1 | NP_001184021.1 | 3 | 283 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gje1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197092.1 | NP_001184021.1 | 3 | 283 |