siRNA Details
| Gene Symbol: Zfpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286936.1 | NP_001273865.1 | 7 | 802 | ||
| XM_006230901.2 | XP_006230963.1 | 7 | 778 | |||
| XM_008760175.1 | XP_008758397.1 | 7 | 766 | |||
| GenBank mRNA | DY546174.1 | - | 5 | 522 | ||
| FQ227385.1 | - | 7 | 838 | |||

Zfpl1
Genomic Map