siRNA Details
| Gene Symbol: Wfdc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253677.1 | NP_001240606.1 | 2 | 138 | ||
| GenBank mRNA | GQ847861.1 | - | 2 | 138 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc11| Gene Symbol: Wfdc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253677.1 | NP_001240606.1 | 2 | 138 | ||
| GenBank mRNA | GQ847861.1 | - | 2 | 138 | ||