siRNA Details
| Gene Symbol: Tmem139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001056308.4 | XP_001056308.1 | 3 | 1536 | ||
| XM_006236395.2 | XP_006236457.1 | 4 | 1584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem139| Gene Symbol: Tmem139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001056308.4 | XP_001056308.1 | 3 | 1536 | ||
| XM_006236395.2 | XP_006236457.1 | 4 | 1584 | |||