siRNA Details
| Gene Symbol: Cfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017147.2 | NP_058843.1 | 4 | 505 | ||
| GenBank mRNA | BC059143.1 | - | 4 | 505 | 6889053 | |
| BC086533.1 | - | 4 | 490 | 7305422 | ||
| FQ212136.1 | - | 3 | 377 | |||
| FQ212794.1 | - | 4 | 504 | |||
| FQ212910.1 | - | 4 | 500 | |||
| FQ213701.1 | - | 4 | 500 | |||
| FQ213897.1 | - | 4 | 529 | |||
| FQ214120.1 | - | 4 | 501 | |||
| FQ214438.1 | - | 4 | 500 | |||
| FQ220384.1 | - | 4 | 516 | |||
| FQ220571.1 | - | 4 | 502 | |||
| FQ226405.1 | - | 4 | 500 | |||
| FQ227487.1 | - | 4 | 502 | |||
| FQ229073.1 | - | 4 | 529 | |||
| FQ229766.1 | - | 4 | 504 | |||
| FQ229833.1 | - | 4 | 500 | |||
| FQ229875.1 | - | 4 | 504 | |||
| FQ230001.1 | - | 4 | 504 | |||
| FQ230091.1 | - | 4 | 501 | |||
| FQ231379.1 | - | 4 | 500 | |||
| FQ233750.1 | - | 4 | 500 | |||
| FQ234162.1 | - | 4 | 504 | |||
| FQ234303.1 | - | 4 | 500 | |||
| X62908.1 | - | 3 | 429 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007687, AK167795, BC046225, AK050651, AK013943, and others.

Cfl1
Genomic Map