siRNA Details
| Gene Symbol: Ttc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750130.3 | XP_003750178.1 | 2 | 392 | ||
| GenBank mRNA | FQ209446.1 | - | 1 | 116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc32| Gene Symbol: Ttc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750130.3 | XP_003750178.1 | 2 | 392 | ||
| GenBank mRNA | FQ209446.1 | - | 1 | 116 | ||