siRNA Details
| Gene Symbol: Sbk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001078116.5 | XP_001078116.3 | 2 | 467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbk3| Gene Symbol: Sbk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001078116.5 | XP_001078116.3 | 2 | 467 | ||