siRNA Details
| Gene Symbol: ADGRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001704.2 | NP_001695.1 | 3 | 847 | ||
| XM_005248752.2 | XP_005248809.1 | 2 | 1133 | |||
| XM_006715534.3 | XP_006715597.1 | 3 | 1269 | |||
| XM_011536010.2 | XP_011534312.1 | 3 | 1000 | |||
| XM_011536011.2 | XP_011534313.1 | 2 | 1133 | |||
| XM_011536012.2 | XP_011534314.1 | 3 | 1269 | |||
| XM_017011140.1 | XP_016866629.1 | 2 | 1097 | |||
| XM_017011142.1 | XP_016866631.1 | 3 | 1269 | |||
| XM_017011143.1 | XP_016866632.1 | 3 | 1269 | |||
| XM_017011144.1 | XP_016866633.1 | 3 | 1269 | |||
| GenBank mRNA | AB005299.1 | - | 2 | 432 | ||
| AB011122.2 | - | 3 | 636 | |||
| AK289696.1 | - | 2 | 448 | |||
| BC111720.1 | - | 1 | 426 | 40027602 | ||
| DB446627.2 | - | 2 | 675 | |||
More Information
Set Membership: |
|

ADGRB3
Genomic Map