siRNA Details
| Gene Symbol: Abracl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099647.1 | NP_001093117.1 | 2 | 111 | ||
| GenBank mRNA | CO569536.1 | - | 2 | 111 | ||
| FQ221434.1 | - | 2 | 174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abracl| Gene Symbol: Abracl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099647.1 | NP_001093117.1 | 2 | 111 | ||
| GenBank mRNA | CO569536.1 | - | 2 | 111 | ||
| FQ221434.1 | - | 2 | 174 | |||