siRNA Details
| Gene Symbol: Gpr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109562.2 | NP_001103032.1 | 4 | 907 | ||
| GenBank mRNA | BC167037.1 | - | 4 | 907 | 7462064 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr137| Gene Symbol: Gpr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109562.2 | NP_001103032.1 | 4 | 907 | ||
| GenBank mRNA | BC167037.1 | - | 4 | 907 | 7462064 | |