siRNA Details
| Gene Symbol: Slc25a54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109640.1 | NP_001103110.1 | 5, 6 | 745 | ||
| XM_008761423.1 | XP_008759645.1 | 5, 6 | 766 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a54| Gene Symbol: Slc25a54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109640.1 | NP_001103110.1 | 5, 6 | 745 | ||
| XM_008761423.1 | XP_008759645.1 | 5, 6 | 766 | |||