siRNA Details
| Gene Symbol: LOC691422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765196.1 | XP_008763418.1 | 1155 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691422| Gene Symbol: LOC691422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765196.1 | XP_008763418.1 | 1155 | |||