siRNA Details
| Gene Symbol: GPRASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099410.1 | NP_001092880.1 | 4 | 4394 | ||
| NM_001099411.1 | NP_001092881.1 | 3 | 4326 | |||
| NM_001184727.1 | NP_001171656.1 | 6 | 4497 | |||
| NM_014710.4 | NP_055525.3 | 5 | 4485 | |||
| XM_017029981.1 | XP_016885470.1 | 4 | 4374 | |||
| XM_017029982.1 | XP_016885471.1 | 4 | 4409 | |||
| GenBank mRNA | AB007903.2 | - | 1 | 3850 | ||
| BC114552.1 | - | 3 | 4130 | 40037190 | ||

GPRASP1
Genomic Map