siRNA Details
| Gene Symbol: Dqx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109407.1 | NP_001102877.1 | 9 | 1645 | ||
| XM_006236717.2 | XP_006236779.1 | 9 | 2135 | |||
| XM_008763030.1 | XP_008761252.1 | 8 | 2320 | |||
| Gene Symbol: LOC103692182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763119.1 | XP_008761341.1 | 201 | |||

Dqx1
Genomic Map