siRNA Details
| Gene Symbol: Kctd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109650.1 | NP_001103120.1 | 1 | 1734 | ||
| XM_006252330.2 | XP_006252392.1 | 2 | 4284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd4| Gene Symbol: Kctd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109650.1 | NP_001103120.1 | 1 | 1734 | ||
| XM_006252330.2 | XP_006252392.1 | 2 | 4284 | |||