siRNA Details
| Gene Symbol: Zfp449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752076.3 | XP_003752124.1 | 5 | 1175 | ||
| XM_008773325.1 | XP_008771547.1 | 4 | 2804 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp449| Gene Symbol: Zfp449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752076.3 | XP_003752124.1 | 5 | 1175 | ||
| XM_008773325.1 | XP_008771547.1 | 4 | 2804 | |||