siRNA Details
Gene Symbol: Zmat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134747.1 | NP_001128219.1 | 5 | 665 | ||
GenBank mRNA | BC168220.1 | - | 5 | 665 | 7320571 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zmat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134747.1 | NP_001128219.1 | 5 | 665 | ||
GenBank mRNA | BC168220.1 | - | 5 | 665 | 7320571 |