siRNA Details
| Gene Symbol: Zmat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134747.1 | NP_001128219.1 | 5 | 627 | ||
| GenBank mRNA | BC168220.1 | - | 5 | 627 | 7320571 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmat4| Gene Symbol: Zmat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134747.1 | NP_001128219.1 | 5 | 627 | ||
| GenBank mRNA | BC168220.1 | - | 5 | 627 | 7320571 | |