siRNA Details
| Gene Symbol: Eno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109908.1 | NP_001103378.1 | 3 | 303 | ||
| NM_012554.3 | NP_036686.2 | 3 | 254 | |||
| XM_006239444.1 | XP_006239506.1 | 3 | 270 | |||
| GenBank mRNA | BC063174.1 | - | 3 | 247 | ||
| BC078896.1 | - | 3 | 303 | 7112523 | ||
| BC081847.1 | - | 3 | 227 | 7189453 | ||
| BC090069.1 | - | 3 | 184 | 7108936 | ||
| BC091572.1 | - | 3 | 224 | 7107492 | ||
| BC097343.1 | - | 3 | 220 | 7461555 | ||
| CB809779.1 | - | 3 | 254 | |||
| FQ214707.1 | - | 3 | 176 | |||
| X02610.1 | - | 3 | 243 | |||

Eno1
Genomic Map