siRNA Details
| Gene Symbol: Meiob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115033.1 | NP_001108505.1 | 11 | 1038 | ||
| XM_006246031.2 | XP_006246093.1 | 11 | 1253 | |||
| XM_008767593.1 | XP_008765815.1 | 11 | 1125 | |||
| GenBank mRNA | BC098928.1 | - | 3 | 902 | ||
| BC158604.1 | - | 11 | 1038 | 7134815 | ||

Meiob
Genomic Map