siRNA Details
| Gene Symbol: Nkx6-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109455.1 | NP_001102925.1 | 2 | 723 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx6-3| Gene Symbol: Nkx6-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109455.1 | NP_001102925.1 | 2 | 723 | ||