siRNA Details
| Gene Symbol: Plp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207601.1 | NP_997484.1 | 4 | 484 | ||
| GenBank mRNA | BC061844.1 | - | 4 | 484 | 5621135 | |
| FQ216622.1 | - | 4 | 520 | |||
| FQ220564.1 | - | 4 | 523 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plp2| Gene Symbol: Plp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207601.1 | NP_997484.1 | 4 | 484 | ||
| GenBank mRNA | BC061844.1 | - | 4 | 484 | 5621135 | |
| FQ216622.1 | - | 4 | 520 | |||
| FQ220564.1 | - | 4 | 523 | |||