siRNA Details
| Gene Symbol: LOC102555023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC088278.1 | - | 2 | 1106 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102555023| Gene Symbol: LOC102555023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC088278.1 | - | 2 | 1106 | ||