siRNA Details
| Gene Symbol: Mea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044286.1 | NP_001037751.1 | 2 | 201 | ||
| XM_006244566.2 | XP_006244628.1 | 1 | 135 | |||
| XM_006244567.2 | XP_006244629.1 | 2 | 256 | |||
| XM_006244568.2 | XP_006244630.1 | 2 | 202 | |||
| XM_006244569.2 | XP_006244631.1 | 2 | 193 | |||
| XM_008766880.1 | XP_008765102.1 | 2 | 301 | |||
| GenBank mRNA | BC089978.1 | - | 2 | 201 | 7317693 | |
| FQ213546.1 | - | 2 | 186 | |||
| FQ223977.1 | - | 2 | 187 | |||

Mea1
Genomic Map