siRNA Details
| Gene Symbol: Cnpy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134710.1 | NP_001128182.1 | 3, 4 | 399 | ||
| GenBank mRNA | BC166762.1 | - | 3, 4 | 399 | 7931606 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnpy3| Gene Symbol: Cnpy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134710.1 | NP_001128182.1 | 3, 4 | 399 | ||
| GenBank mRNA | BC166762.1 | - | 3, 4 | 399 | 7931606 | |