siRNA Details
| Gene Symbol: SLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304743.1 | NP_001291672.1 | 9, 10 | 2902 | ||
| NM_014720.3 | NP_055535.2 | 9, 10 | 2902 | |||
| GenBank mRNA | AB002804.1 | - | 9, 10 | 2573 | ||
| AF273048.1 | - | 9, 10 | 2375 | |||
| AK302855.1 | - | 9, 10 | 2335 | |||
| BC111565.1 | - | 9, 10 | 2372 | 40009070 | ||
| D86959.1 | - | 9, 10 | 2852 | |||

SLK
Genomic Map