siRNA Details
| Gene Symbol: Ears2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159493.1 | NP_001152965.1 | 3 | 467 | ||
| Gene Symbol: Ears2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760356.1 | XP_008758578.1 | 1 | 400 | ||

Ears2
Genomic Map